Merck
  • The electrochemical detection of tagged nanoparticles via particle-electrode collisions: nanoelectroanalysis beyond immobilisation.
The electrochemical detection of tagged nanoparticles via particle-electrode collisions: nanoelectroanalysis beyond immobilisation.
Chemical communications (Cambridge, England) (2012-01-27)
Yi-Ge Zhou, Neil V Rees, Richard G Compton
ABSTRACT
The use of particle-impact coulometry in identifying and quantifying nanoparticles tagged (or labelled) with electroactive molecules is demonstrated via the detection of 1,4-nitrothiophenol-tagged silver nanoparticles in aqueous dispersion at potentials more negative than -0.17 V (vs. Ag/AgCl, the reduction potential of nitrothiophenol) via monitoring of particle-electrode collisions.
Product Description
Supelco
Thiophenol, analytical standard
Sigma-Aldrich
Sodium thiophenolate, technical grade, 90%
Sigma-Aldrich
Thiophenol, 97%
Sigma-Aldrich
Thiophenol, ≥99%