MilliporeSigma
Dexrazoxane treatment of doxorubicin extravasation injury in four dogs.
Journal of the American Veterinary Medical Association (2012-01-20)
Rachel O Venable, Corey F Saba, Melissa M Endicott, Nicole C Northrup
ABSTRACT
4 dogs were treated with dexrazoxane for known or suspected doxorubicin extravasation. Records were retrospectively reviewed. Doses and number of doses of dexrazoxane were variable. Dexrazoxane was administered within 2 hours after known extravasation in 3 dogs and 48 hours after suspected extravasation in 1 dog. Additional medical treatments included tissue cooling in all dogs, topically administered dimethyl sulfoxide ointment in 3, and orally administered piroxicam in 1. Mild erythema and edema at the extravasation site developed within 1 to 6 days after extravasation in the 3 dogs that received dexrazoxane within 2 hours after extravasation. Extensive tissue necrosis occurred in the dog treated 48 hours after suspected extravasation. Only the dog with severe tissue necrosis required surgical intervention. Lesions in the other 3 dogs resolved with medical management alone. All dogs survived the event. To date, use of dexrazoxane in the management of doxorubicin extravasation has not been reported in dogs. Treatment was successful in 3 of 4 patients. The most effective dosage and timing of administration are unknown; however, there is evidence to suggest that administration within 6 hours after the event is warranted. Further studies are needed to confirm efficacy and to optimize use of this drug in the prevention and treatment of anthracycline extravasation injury in veterinary patients.
Sigma-Aldrich
Dexrazoxane, ≥95% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Razoxane, >98% (HPLC)