Merck
  • A state-wide survey in Oregon (USA) of trace metals and organic chemicals in municipal effluent.
A state-wide survey in Oregon (USA) of trace metals and organic chemicals in municipal effluent.
The Science of the total environment (2012-01-17)
Bruce K Hope, Lori Pillsbury, Brian Boling
ABSTRACT
Oregon's Senate Bill 737, enacted in 2007, required the state's 52 largest municipal wastewater treatment plants (WWTP) and water pollution control facilities (WPCF) to collect effluent samples in 2010 and analyze them for persistent organic pollutants. These facilities are located state-wide and represent a variety of treatment types, service population sizes, geographic areas, and flow conditions. Of the 406 chemicals ultimately analyzed, 114 were detected above the level of quantification (LOQ) in at least one sample. Few persistent pollutants were found possibly because of their diversion from effluent via sorption to sludge (solids phase) or high LOQs for certain chemicals. Several pesticides, as well as benzene and phenol degradation products, all previously unreported in effluent, were detected. Ten polychlorinated biphenyls (PCB) congeners were present at low concentrations in ≤ 10 samples, while polychlorinated naphthalenes and dioxins/furans were not detected at all. Twenty-one polybrominated diphenyl ether (PBDE) congeners were found, nine of which have been reported in Osprey eggs in Oregon and Washington. Methylmercury was present in 65% of samples, with average and maximum concentrations of 0.18 and 1.36 ng/L, respectively. Although they are generally assumed to be innocuous by-products of sewage treatment, additional research is needed on potential impacts to aquatic ecosystems of high loadings of coprostanol and cholesterol. These results suggest that effluent, rather than just receiving waters, should itself be analyzed for a wide range of contaminants in order to understand how upstream sources, conveyed through WWTPs and WPCFs, could be impacting aquatic ecosystems.
Sigma-Aldrich
5α-Cholestan-3β-ol, ≥95%
Sigma-Aldrich
Coprostan-3-ol, ≥98%
Sigma-Aldrich
5β-Cholestan-3α-ol, ≥95%