Merck
  • Effect of ionic liquid on diffusion in P123 gel: fluorescence correlation spectroscopy.
Effect of ionic liquid on diffusion in P123 gel: fluorescence correlation spectroscopy.
Chemphyschem : a European journal of chemical physics and physical chemistry (2012-01-10)
Amit Kumar Mandal, Supratik Sen Mojumdar, Atanu Kumar Das, Kankan Bhattacharyya
ABSTRACT
The effect of two room-temperature ionic liquids (RTILs) on the diffusion of three fluorescent dyes in the gel phase of a triblock copolymer, (PEO)(20)-(PPO)(70)-(PEO)(20) [Pluronic P123; poly ethylene oxide (PEO), poly propylene oxide (PPO)], was studied by using fluorescence correlation spectroscopy (FCS). We used three dyes, 4-(dicyanomethylene)-2-methyl-6-(4-dimethylaminostyryl)-4H-pyran (DCM), coumarin 480 (C480), and coumarin 343 (C343). By field-emission scanning electron microscopy (FESEM), it was observed that the macroscopic structure of the P123 gel remained unaffected upon addition of RTIL. In the absence of RTIL, the diffusion coefficient (D(t)) of the hydrophobic dye DCM (1 μm(2) s(-1) at the core) is smaller than that of the other two hydrophilic dyes (7 μm(2) s(-1) for C480 and C343). On addition of RTIL, the D(t) values of all of the dyes increase, indicating a decrease in local viscosity (η(eff)). The η(eff) of the core of the RTIL-P123 gel estimated from the D(t) of DCM is lower than that of both the P123 gel (at the core η=90 cP) and RTIL (η=110 cP). It is shown that the RTIL affects the structure of the gel by modifying the size of the micellar aggregates and by penetrating the core.
Sigma-Aldrich
Poloxamer 188, micronized
Sigma-Aldrich
Poloxamer 188 solution, 10%, sterile-filtered, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,000
Sigma-Aldrich
4-(Dicyanomethylene)-2-methyl-6-(4-dimethylaminostyryl)-4H-pyran, Dye content 98 %
Sigma-Aldrich
Poloxamer 407, micronized
Sigma-Aldrich
Synperonic® PE/P84
Sigma-Aldrich
Poloxamer 407, purified, non-ionic
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,100
Sigma-Aldrich
Synperonic® PE P105, surfactant
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~3,300
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~2,000
Sigma-Aldrich
Poloxamer 188, solid
Sigma-Aldrich
Synperonic® F 108, surfactant, non-ionic
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~14,600
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,900
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~8,400
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,900
Sigma-Aldrich
Poloxamer 188
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~4,400
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~5,800
Sigma-Aldrich
Poloxamer 407, oxyethylene 71.5-74.9 %