Merck
  • Development of udenafil-loaded microemulsions for intranasal delivery: in vitro and in vivo evaluations.
Development of udenafil-loaded microemulsions for intranasal delivery: in vitro and in vivo evaluations.
International journal of pharmaceutics (2012-01-03)
Hyun-Jong Cho, Wan-Sung Ku, Ubonvan Termsarasab, Insoo Yoon, Chung-Wook Chung, Hyun Tae Moon, Dae-Duk Kim
ABSTRACT
To achieve rapid onset of action and improved bioavailability of udenafil, a microemulsion system was developed for its intranasal delivery. Phase behavior, particle size, transmission electron microscope (TEM) images, and the drug solubilization capacity of the microemulsion were investigated. A single isotropic region was found in pseudo-ternary phase diagrams developed at various ratios with CapMul MCM L8 as an oil, Labrasol as a surfactant, and Transcutol or its mixture with ethanol (1:0.25, v/v) as a cosurfactant. Optimized microemulsion formulations with a mean diameter of 120-154 nm achieved enhanced solubility of udenafil (>10mg/ml) compared with its aqueous solubility (0.02 mg/ml). An in vitro permeation study was performed in human nasal epithelial (HNE) cell monolayers cultured by the air-liquid interface (ALI) method, and the permeated amounts of udenafil increased up to 3.41-fold versus that of pure udenafil. According to the results of an in vivo pharmacokinetic study in rats, intranasal administration of udenafil-loaded microemulsion had a shorter T(max) value (1 min) compared with oral administration and improved bioavailability (85.71%) compared with oral and intranasal (solution) administration. The microemulsion system developed for intranasal administration may be a promising delivery system of udenafil, with a rapid onset of action and improved bioavailability.
Sigma-Aldrich
Di(ethylene glycol) ethyl ether, ≥99%
Sigma-Aldrich
Diethylene glycol monoethyl ether, ReagentPlus®, 99%