Merck
  • Emerging pollutants in sewage, surface and drinking water in Galicia (NW Spain).
Emerging pollutants in sewage, surface and drinking water in Galicia (NW Spain).
Chemosphere (2011-12-23)
Rosario Rodil, José Benito Quintana, Estefanía Concha-Graña, Purificación López-Mahía, Soledad Muniategui-Lorenzo, Darío Prada-Rodríguez
ABSTRACT
A monitoring programme was carried out on wastewater, surface and drinking water on the NW area of Spain during the four seasons of a year period (November 2007-September 2008). This study covered a series of emerging pollutants of different classes, including pharmaceuticals, neutral and acidic organophosphorus flame retardant/plasticizers (OPs), triclosan, phenoxy-herbicides, insect repellents and UV filters. From the total set of 53 compounds, 19 were found in raw wastewater with median concentrations higher than 0.1 μg L(-1). Among them, salicylic acid, ibuprofen and the UV filter benzophenone-4 (BP-4) were the most concentrated, exceeding the 1 μg L(-1) median value. Subsequently, 11 of these contaminants are not efficiently enough removed in the small WWTPs tested and their median concentrations in effluents still surpassed the 0.1 μg L(-1), so that they can spread through surface water. These chemicals are the pharmaceuticals naproxen, diclofenac and atenolol; the OPs tri(2-chloroethyl) phosphate (TCEP), tri(chloropropyl) phosphate (TCPP), tri-n-butyl phosphate (TnBP), diphenyl phosphate (DPhP) and diethylhexyl phosphate (DEHP); and the sulphonate UV filters BP-4 and 2-phenylbenzimidazole-5-sulphonic acid (PBSA). These OPs were then the dominant emerging pollutants occurring in surface and drinking water, where they are detected in the 20-200 ng L(-1) range. Pharmaceuticals and UV filters are typically below the 10 ng L(-1) level. Finally, herbicides were only detected in the last sampling campaign under the 100 ng L(-1) drinking water European Union limit.
Product Description
Supelco
Sulisobenzone, analytical standard
Sigma-Aldrich
Tributyl phosphate, 97%
Sigma-Aldrich
Tributyl phosphate, puriss., ≥99.0% (GC)
Sigma-Aldrich
5-Benzoyl-4-hydroxy-2-methoxybenzenesulfonic acid, ≥97.0% (HPLC)
Sigma-Aldrich
Tributyl phosphate, ≥99%
Supelco
Tributyl phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Tri-n-butyl phosphate, European Pharmacopoeia (EP) Reference Standard
Supelco
Tributyl phosphate, for extraction analysis, ≥99.0% (GC)