Merck
  • Multiplexing interactions to control antibiotic release from cyclodextrin hydrogels.
Multiplexing interactions to control antibiotic release from cyclodextrin hydrogels.
Macromolecular bioscience (2011-12-15)
Thimma R Thatiparti, Nicole Averell, Derek Overstreet, Horst A von Recum
ABSTRACT
A new strategy for affinity-based drug delivery by modification of the drug rather than modification of the device is presented. Rifampin is modified to contain either one or two PEG-adamantane arms, and the drug release properties of dimeric coumermycin are compared to novobiocin with only one biding domain. The drugs are loaded into affinity-based and diffusion-only delivery platforms, the loading efficiency is calculated, and the release kinetics is determined in vitro. The presence of additional binding domains prolongs the release of antibiotics. Release rates differ little between modified and unmodified drug from the diffusion-only system. The results demonstrate the feasibility of custom-tuning drug delivery by multiplexing interactions with an affinity-based polymer platform.
Sigma-Aldrich
Novobiocin sodium salt, ≥90% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-1719793-50MG50 mg
Ships in 14 days - from Aldrich Partner.
HUF 239.894,00
EN300-1719793-100MG100 mg
Ships in 14 days - from Aldrich Partner.
HUF 308.560,00
EN300-1719793-250MG250 mg
Ships in 14 days - from Aldrich Partner.
HUF 432.554,00
EN300-1719793-500MG500 mg
Ships in 14 days - from Aldrich Partner.
HUF 672.638,00
EN300-1719793-1G1 g
Ships in 14 days - from Aldrich Partner.
HUF 857.394,00
EN300-1719793-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
HUF 1.664.096,00
EN300-1719793-5G5 g
Ships in 14 days - from Aldrich Partner.
HUF 2.453.508,00
EN300-1719793-10G10 g
Ships in 14 days - from Aldrich Partner.
HUF 3.630.710,00
Sigma-Aldrich
Novobiocin sodium, meets USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-1719793-50MG50 mg
Ships in 14 days - from Aldrich Partner.
HUF 239.894,00
EN300-1719793-100MG100 mg
Ships in 14 days - from Aldrich Partner.
HUF 308.560,00
EN300-1719793-250MG250 mg
Ships in 14 days - from Aldrich Partner.
HUF 432.554,00
EN300-1719793-500MG500 mg
Ships in 14 days - from Aldrich Partner.
HUF 672.638,00
EN300-1719793-1G1 g
Ships in 14 days - from Aldrich Partner.
HUF 857.394,00
EN300-1719793-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
HUF 1.664.096,00
EN300-1719793-5G5 g
Ships in 14 days - from Aldrich Partner.
HUF 2.453.508,00
EN300-1719793-10G10 g
Ships in 14 days - from Aldrich Partner.
HUF 3.630.710,00
Supelco
Novobiocin sodium salt, VETRANAL®, analytical standard
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-1719793-50MG50 mg
Ships in 14 days - from Aldrich Partner.
HUF 239.894,00
EN300-1719793-100MG100 mg
Ships in 14 days - from Aldrich Partner.
HUF 308.560,00
EN300-1719793-250MG250 mg
Ships in 14 days - from Aldrich Partner.
HUF 432.554,00
EN300-1719793-500MG500 mg
Ships in 14 days - from Aldrich Partner.
HUF 672.638,00
EN300-1719793-1G1 g
Ships in 14 days - from Aldrich Partner.
HUF 857.394,00
EN300-1719793-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
HUF 1.664.096,00
EN300-1719793-5G5 g
Ships in 14 days - from Aldrich Partner.
HUF 2.453.508,00
EN300-1719793-10G10 g
Ships in 14 days - from Aldrich Partner.
HUF 3.630.710,00