Merck
  • Production of p-acetaminophenol by whole-cell catalysis using Escherichia coli overexpressing bacterial aryl acylamidase.
Production of p-acetaminophenol by whole-cell catalysis using Escherichia coli overexpressing bacterial aryl acylamidase.
Biotechnology letters (2011-12-02)
Hyeok-Jin Ko, Won-Gi Bang, Kyoung Heon Kim, In-Geol Choi
ABSTRACT
Aryl acylamidase (EC 3.5.1.13, AAA) acts on the amide bond between aryl and acyl groups. Whole cells of Escherichia coli overexpressing a novel bacterial AAA synthesized p-acetaminophenol (p-AAP) from p-aminophenol (p-AP, aryl compound) and acetate (acyl donor). Optimum conditions were pH 5.5 and 35°C with 100 mM p-AP and 600 mM sodium acetate in 100 mM sodium phosphate buffer including 1% (v/v) Triton X-100 for 60 h. 13.1 g p-AAP l(-1) was produced with a conversion yield of 87%.
Product Description
Supelco
4-Aminophenol, PESTANAL®, analytical standard
Supelco
4-Aminophenol (Acetaminophen Related Compound K) (Paracetamol Impurity K), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Aminophenol, ≥98%