Merck
  • Stepwise fluorescence changes of quantum dots: single-molecule spectroscopic studies on the properties of turn-on quantum dots.
Stepwise fluorescence changes of quantum dots: single-molecule spectroscopic studies on the properties of turn-on quantum dots.
Chemical communications (Cambridge, England) (2011-11-26)
Yea Seul Kim, Min Young Kim, Jae Kyu Song, Tae Jung Kim, Young Dong Kim, Sang Soo Hah
ABSTRACT
Single-molecule spectroscopy of turn-on quantum dots induced by NADPH-dependent biocatalyzed transformations reveals that the fluorescence intensities of quantum dots functionalized with Nile Blue are stepwisely and reversibly changed in the presence of NADPH.
Sigma-Aldrich
Nile Blue A perchlorate, Dye content 95 %
Sigma-Aldrich
Nile Blue chloride, Dye content 85 %
Sigma-Aldrich
Nile Blue A, certified by the Biological Stain Commission
Sigma-Aldrich
Nile Blue A, Dye content ≥75 %