Merck
  • Effect of cation disorder on structural, magnetic and dielectric properties of La2MnCoO6 double perovskite.
Effect of cation disorder on structural, magnetic and dielectric properties of La2MnCoO6 double perovskite.
Journal of physics. Condensed matter : an Institute of Physics journal (2011-11-24)
A J Barón-González, C Frontera, J L García-Muñoz, B Rivas-Murias, J Blasco
ABSTRACT
The origin of dielectric anomalies and magnetodielectric response of La(2)MnCoO(6) has been investigated by means of ultra-high resolution synchrotron x-ray powder diffraction, neutron powder diffraction, resistivity, magnetization and dielectric measurements. The study has been performed on two different bulk samples presenting different degrees of Mn/Co order: 95 and 74%. Beside the well-known influence on magnetic properties, our results show that the main effect of disorder lies on the electrical resistivity. Bond distances clearly show Mn(4+)/Co(2+) valence states in the well-ordered sample, while for the disordered one this picture still holds. AC resistivity data show dielectric anomalies and a small magnetodielectric effect, but impedance complex plane analyses prove that these phenomena appear at the frequency-temperature region where extrinsic effects dominate the dielectric response.
Sigma-Aldrich
Lanthanum(III) oxide, 99.99% trace metals basis
Supelco
Lanthanum(III) oxide, suitable for AAS, ≥99.9%
Sigma-Aldrich
Lanthanum(III) oxide, 99.999% trace metals basis
Sigma-Aldrich
Lanthanum(III) oxide, nanopowder, <100 nm particle size (TEM), 99% trace metals basis
Sigma-Aldrich
Lanthanum(III) oxide, ≥99.9%