Merck
  • RGD-based active targeting of novel polycation liposomes bearing siRNA for cancer treatment.
RGD-based active targeting of novel polycation liposomes bearing siRNA for cancer treatment.
Journal of controlled release : official journal of the Controlled Release Society (2011-10-25)
Norihito Yonenaga, Eriya Kenjo, Tomohiro Asai, Atsushi Tsuruta, Kosuke Shimizu, Takehisa Dewa, Mamoru Nango, Naoto Oku
ABSTRACT
For the purpose of systemic delivery of siRNA, we previously developed polycation liposomes (PCLs) containing dicetylphosphate-tetraethylenepentamine (DCP-TEPA) as an effective siRNA carrier. In the present study, to endow these PCLs (TEPA-PCL) actively target cancer cells and angiogenic vessels, we decorated the PCLs with cyclic RGD, by using cyclic RGD-grafted distearoylphosphatidylethanolamine-polyethylene glycol (DSPE-PEG), and investigated the usefulness of this type of carrier (RGD-PEG-PCL) for active targeting. Firstly, the gene-silencing efficacy of siRNA for luciferase (siLuc2) formulated in RGD-PEG-PCL (RGD-PEG-PCL/siLuc2) was examined in vitro by using B16F10-luc2 murine melanoma cells stably expressing the luciferase 2 gene, where the siRNA was grafted with cholesterol at the 3'-end of the sense strand (siRNA-C) for the stable association of the siRNA with the PCL. RGD-PEG-PCL/siLuc2 showed high knockdown efficiency compared with siLuc2 formulated in PEGylated TEPA-PCL without cyclic RGD (PEG-PCL). Next, the gene-silencing efficacy of RGD-PEG-PCL/siLuc2 was examined in vivo by use of B16F10-luc2 lung metastatic model mice. The intravenous injection of RGD-PEG-PCL/siLuc2 showed high knockdown efficiency against metastatic B16F10-luc2 tumors in the lungs of the mice, as assessed with an in vivo imaging system. These data strongly suggest that systemic and active targeting siRNA delivery using RGD-PEG-PCL is useful for cancer RNAi therapy.
Sigma-Aldrich
Dihexadecyl phosphate
Sigma-Aldrich
Tetraethylenepentamine, technical grade