Merck
  • Simultaneous analysis of pesticides from different chemical classes by using a derivatisation step and gas chromatography-mass spectrometry.
Simultaneous analysis of pesticides from different chemical classes by using a derivatisation step and gas chromatography-mass spectrometry.
Journal of chromatography. A (2011-10-04)
Caroline Raeppel, Marie Nief, Marie Fabritius, Lucie Racault, Brice M Appenzeller, Maurice Millet
ABSTRACT
This work presents a new method to analyse simultaneously by GC-MS 31 pesticides from different chemical classes (2,4 D, 2,4 MCPA, alphacypermethrin, bifenthrin, bromoxynil, buprofezin, carbaryl, carbofuran, clopyralid, cyprodinil, deltamethrin dicamba, dichlobenil, dichlorprop, diflufenican, diuron, fenoxaprop, flazasulfuron, fluroxypyr, ioxynil, isoxaben, mecoprop-P, myclobutanil, oryzalin, oxadiazon, picloram, tau-fluvalinate tebuconazole, triclopyr, trifluralin and trinexapac-p-ethyl). This GC-MS method will be applied to the analysis of passive samplers (Tenax(®) tubes and SPME fiber) used for the evaluation of the indoor and outdoor atmospheric contamination by non-agricultural pesticides. The method involves a derivatisation step for thermo-labile or polar pesticides. Different agents were tested and MtBSTFA (N-(t-butyldimethylsilyl)-N-methyltrifluoroacetamide), a sylilation agent producing very specific fragments [M-57], was retained. However, diuron could not be derivatised and the isocyanate product was used for identification and quantification. Pesticides which did not need a derivatisation step were not affected by the presence of the derivatisation agent and they could easily be analysed in mixture with derivatised pesticides. The method can be coupled to a thermal-desorption unit or to SPME extraction for a multiresidue analysis of various pesticides in atmospheric samples.
Product Description
Supelco
N-Methyl-bis(trifluoroacetamide), LiChropur, ≥97.0% (GC)
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide, >97%
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, ≥95%
Supelco
MTBSTFA (with 1% t-BDMCS), (with 1% t-BDMCS), ampule of 5 × 1 mL, analytical standard, Cerilliant®
Supelco
N-Methyl-bis(trifluoroacetamide), derivatization grade (GC derivatization), LiChropur, ≥99.0% (GC)