Merck
  • One-pot synthesis of an organic-inorganic hybrid affinity monolithic column for specific capture of glycoproteins.
One-pot synthesis of an organic-inorganic hybrid affinity monolithic column for specific capture of glycoproteins.
Chemical communications (Cambridge, England) (2011-07-30)
Zian Lin, Jilei Pang, Huanghao Yang, Zongwei Cai, Lan Zhang, Guonan Chen
ABSTRACT
An inorganic-organic hybrid affinity monolithic column was synthesized by a novel "one-pot" approach. The resulting hybrid affinity monoliths have potential applications in specific recognition and enrichment of glycoproteins.
Sigma-Aldrich
Tetramethyl orthosilicate, ≥99%
Sigma-Aldrich
Tetramethyl orthosilicate, 98%
Sigma-Aldrich
Tetramethyl orthosilicate, purum, ≥98.0% (GC)