Merck
Diversified applications of chemically modified 1,2-polybutadiene.
Macromolecular rapid communications (2011-06-29)
Jiayin Yuan, Niels ten Brummelhuis, Mathias Junginger, Zailai Xie, Yan Lu, Andreas Taubert, Helmut Schlaad
ABSTRACT
Commercially available 1,2-PB was transformed into a well-defined reactive intermediate by quantitative bromination. The brominated polymer was used as a polyfunctional macroinitiator for the cationic ring-opening polymerization of 2-ethyl-2-oxazoline to yield a water-soluble brush polymer. Nucleophilic substitution of bromide by 1-methyl imidazole resulted in the formation of polyelectrolyte copolymers consisting of mixed units of imidazolium, bromo, and double bond. These copolymers, which were soluble in water without forming aggregates, were used as stabilizers in the heterophase polymerization of styrene and were also studied for their ionic conducting properties.
Sigma-Aldrich
Polybutadiene, average Mn ~5,000
Sigma-Aldrich
Polybutadiene, predominantly 1,2-addition, approx. 90% 1,2-vinyl
Sigma-Aldrich
Polybutadiene, cis, average Mw 200,000-300,000