Merck
  • Non-chromatographic screening procedure for arsenic speciation analysis in fish-based baby foods by using electrothermal atomic absorption spectrometry.
Non-chromatographic screening procedure for arsenic speciation analysis in fish-based baby foods by using electrothermal atomic absorption spectrometry.
Analytica chimica acta (2011-06-28)
Ignacio López-García, Marisol Briceño, Manuel Hernández-Córdoba
ABSTRACT
A procedure for the speciation analysis of arsenic in fish-based baby foods is presented. Inorganic arsenic, methylarsonic acid (MA), dimethylarsinic acid (DMA) and arsenobetaine (AB) were determined by electrothermal atomic absorption spectrometry (ETAAS) using suspensions prepared in a 0.01 mol L(-1) tetramethylammonium hydroxide (TMAH) solution. Speciation is based on the use of three different chemically modified ETAAS atomizers to obtain the analytical signals. Using a palladium salt as the chemical modifier, the signal corresponding to the total arsenic concentration is obtained. When palladium is replaced by Ce(IV), the signal is solely due to inorganic arsenic (III and V)+MA. If no signal is obtained in this latter case, it is possible to distinguish between DMA and AB using a zirconium coated atomizer. The signal obtained in this way is due solely to DMA, and the concentration of AB can be obtained by the difference with the total arsenic content. Determinations by ETAAS require the use of the standard additions method. The limits of detection for the determination of AB, DMA and inorganic arsenic (+MA) are 15, 25 and 50 ng g(-1) expressed as arsenic, respectively. These detection limits are good enough for the procedure to be appropriate for the rapid determination of these compounds, avoiding extraction processes and/or chromatographic separations. Data for commercial samples, as well as for four standard reference materials, are given.
Sigma-Aldrich
Tetramethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)
Supelco
Tetramethylammonium sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Tetramethylammonium chloride, reagent grade, ≥98%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium fluoride, 97%
Supelco
Tetramethylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium iodide, 99%
Supelco
Tetramethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium bromide, 98%
Sigma-Aldrich
Tetramethylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
Sigma-Aldrich
Arsenobetaine, purum p.a., ≥95.0% (NMR)
Sigma-Aldrich
Tetramethylammonium chloride solution, for molecular biology
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥97%