- Hypolipidemic activity of Symplocos cochinchinensis S. Moore leaves in hyperlipidemic rats.
Hypolipidemic activity of Symplocos cochinchinensis S. Moore leaves in hyperlipidemic rats.
The hypolipidemic activity of Symplocos cochinchinensis S. Moore leaves was studied in Triton WR-1339- and high fat diet-induced hyperlipidemic rats. In Triton WR-1339-induced hyperlipidemic rats, the hexane extract (250 and 500 mg/kg) exerted a significant (P < 0.01) lipid-lowering effect compared to ethyl acetate and methanol extracts, as assessed by the reversal of the plasma levels of total cholesterol (TC), triglycerides (TG), high-density lipoprotein cholesterol (HDL-C), and low-density lipoprotein cholesterol (LDL-C). In high fat diet-fed hyperlipidemic rats, the hexane extract (250 and 500 mg/kg) caused the lowering of lipid levels in the plasma and liver. The hypolipidemic activity of S. cochinchinensis leaves was compared with fenofibrate, a known lipid-lowering drug, in both models.