Merck
  • Ability to use the diazo dye, C.I. Acid Black 1 as a nitrogen source by the marine cyanobacterium Oscillatoria curviceps BDU92191.
Ability to use the diazo dye, C.I. Acid Black 1 as a nitrogen source by the marine cyanobacterium Oscillatoria curviceps BDU92191.
Bioresource technology (2011-05-17)
Balakrishnan Priya, Lakshmanan Uma, Abdul Khaleel Ahamed, Gopalakrishnan Subramanian, Dharmar Prabaharan
ABSTRACT
Ten different strains of marine cyanobacteria were tested for their ability to decolourise and degrade a recalcitrant diazo dye, C.I. Acid Black 1. Of them, Oscillatoria curvicepsBDU92191 was able to grow up to a tested concentration of 500 mG L(-1). The organism degraded 84% of the dye at 100 mG L(-1) in 8 days in a medium free of combined nitrogen. The dye degrading ability is attributed to the activities of the enzymes: laccase, polyphenol oxidase and azoreductase. The absence of the doublet amine peak in addition to the overall reduction of absorption in the IR spectra confirmed the mineralisation of the tested azo dye. The nitrogen assimilating enzyme studies along with nitrogenase assay strongly suggested the ability of the non-heterocystous, filamentous marine cyanobacterium, O. curvicepsBDU92191 to use C.I. Acid Black 1 as a nitrogen source in an oligotrophic environment.
Sigma-Aldrich
Naphthol Blue Black, Dye content 80 %
Sigma-Aldrich
Naphthol Blue Black, Dye content ~50 %
Sigma-Aldrich
Naphthol Blue Black, BioReagent, suitable for electrophoresis
Sigma-Aldrich
Amido Black Staining Solution 2X, electrophoresis reagent