Merck
  • The novel δ opioid receptor agonist KNT-127 produces antidepressant-like and antinociceptive effects in mice without producing convulsions.
The novel δ opioid receptor agonist KNT-127 produces antidepressant-like and antinociceptive effects in mice without producing convulsions.
Behavioural brain research (2011-05-14)
Akiyoshi Saitoh, Azusa Sugiyama, Toru Nemoto, Hideaki Fujii, Keiji Wada, Jun-Ichiro Oka, Hiroshi Nagase, Mitsuhiko Yamada
ABSTRACT
We previously reported that the δ opioid receptor (DOP) agonists SNC80 and TAN-67 produce potent antidepressant-like and antinociceptive effects in rodents. However, SNC80 produced convulsive effects. Recently, we succeeded in synthesizing a novel DOP agonist called KNT-127. The present study examined the convulsive, antidepressant-like, and antinociceptive effects of KNT-127 in mice. In contrast to SNC80, KNT-127 produced no convulsions at doses of up to 100mg/kg. In mice subjected to the forced swim test, a screening model for antidepressants, KNT-127 (1mg/kg, s.c.) significantly decreased the duration of immobility and increased the duration of swimming without influencing spontaneous locomotor activity. These behavioral changes were similar to that observed for the tricyclic antidepressant imipramine (6mg/kg). The antidepressant-like effect of KNT-127 in mice was antagonized by pretreatment with naltrindole (NTI), a selective DOP antagonist, or naltriben, a putative DOP(2) subtype antagonist. In addition, KNT-127 (3mg/kg, s.c.) significantly reduced the number of acetic acid-induced abdominal constrictions and the duration of licking time, respectively, in mice subjected to a writhing test and a formalin test. These antinociceptive effects were antagonized by pretreatment with either NTI or 7-benzylidenenaltrexone, a putative DOP(1) subtype antagonist. We propose that KNT-127 should be considered as a candidate compound for the development of DOP-based antidepressants and/or analgesics that lack convulsive effects.
Sigma-Aldrich
SNC80, ≥98% (HPLC), solid
Pricing and availability is not currently available.