Merck
  • Thia-, aza-, and selena[3.3.1]bicyclononane dichlorides: rates vs internal nucleophile in anchimeric assistance.
Thia-, aza-, and selena[3.3.1]bicyclononane dichlorides: rates vs internal nucleophile in anchimeric assistance.
The Journal of organic chemistry (2011-05-07)
Adrian A Accurso, So-Hye Cho, Asmarah Amin, Vladimir A Potapov, Svetlana V Amosova, M G Finn
ABSTRACT
Sulfur-, selenium-, and nitrogen-containing compounds bearing leaving groups in the β-position undergo facile substitution chemistry enabled by anchimeric assistance. Here we provide direct comparisons between such systems in the rigid bicyclo[3.3.1]nonane framework easily derived from 1,5-cyclooctadiene. For a series of dichloride electrophiles of this type, the relative reactivities were found to be Se ≫ (alkyl)N > S ≥ (propargyl)N > (phenyl)N, with the reaction rates at the two extremes differing by more than 3 orders of magnitude. For the N-alkyl case, substitution rates were largely independent of the trapping nucleophile but were strongly dependent on solvent, showing that the process is controlled by the formation of the high-energy three-membered cationic intermediate.
Product Description
Supelco
Nonane, analytical standard
Sigma-Aldrich
Nonane, ReagentPlus®, 99%
Sigma-Aldrich
Nonane, anhydrous, ≥99%