Merck
  • Enhanced immobilization of acidic proteins in the apatite layer via electrostatic interactions in a supersaturated calcium phosphate solution.
Enhanced immobilization of acidic proteins in the apatite layer via electrostatic interactions in a supersaturated calcium phosphate solution.
Acta biomaterialia (2011-05-04)
Ayako Oyane, Teruhisa Ootsuka, Kazumi Hayama, Yu Sogo, Atsuo Ito
ABSTRACT
Artificial materials coated with a protein-apatite composite layer have great potential in clinical applications as a third generation biomaterial. Such composite materials can be prepared by immersing a surface modified substrate into a supersaturated calcium phosphate solution (CP solution: 142 mM NaCl, 3.75 mM CaCl(2), 1.5mM K(2)HPO(4)·3H(2)O, buffered at pH 7.4 at 25 °C with tris(hydroxymethyl)aminomethane and HCl) supplemented with a protein. In the present study proteins of various molecular weights (MW) and isoelectric points (pI) were used to form a protein-apatite composite layer on a polymeric material to determine how the molecular properties of the protein affect the efficiency of protein immobilization (i.e. the amount of immobilized protein in the apatite layer as a percentage of the total amount of protein in solution). The results indicated that the efficiency of protein immobilization did not correlate with the MW of the protein. In contrast, the efficiency of protein immobilization was strongly related to the pI of the protein. As the pI decreased the efficiency of protein immobilization increased due to the high adsorption affinity of negatively charged acidic proteins for positively charged apatite crystals and/or apatite precursors in the CP solution. Thus, the use of acidic rather than basic proteins improves the immobilization efficiency in the present coating process.
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 12 wt. %, melt index 8 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 18 wt. %, melt index 8 g/10 min (190°C/2.16kg), contains 200-900 ppm BHT as inhibitor
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 40 wt. %, melt index (41-63 dg/min (190°C/2.16kg)), contains 190-910 ppm inhibitor