Merck
  • Product selectivity control induced by using liquid-liquid parallel laminar flow in a microreactor.
Product selectivity control induced by using liquid-liquid parallel laminar flow in a microreactor.
Organic & biomolecular chemistry (2011-04-13)
Fumihiro Amemiya, Hideyuki Matsumoto, Keishi Fuse, Tsuneo Kashiwagi, Chiaki Kuroda, Toshio Fuchigami, Mahito Atobe
ABSTRACT
Product selectivity control based on a liquid-liquid parallel laminar flow has been successfully demonstrated by using a microreactor. Our electrochemical microreactor system enables regioselective cross-coupling reaction of aldehyde with allylic chloride via chemoselective cathodic reduction of substrate by the combined use of suitable flow mode and corresponding cathode material. The formation of liquid-liquid parallel laminar flow in the microreactor was supported by the estimation of benzaldehyde diffusion coefficient and computational fluid dynamics simulation. The diffusion coefficient for benzaldehyde in Bu(4)NClO(4)-HMPA medium was determined to be 1.32 × 10(-7) cm(2) s(-1) by electrochemical measurements, and the flow simulation using this value revealed the formation of clear concentration gradient of benzaldehyde in the microreactor channel over a specific channel length. In addition, the necessity of the liquid-liquid parallel laminar flow was confirmed by flow mode experiments.
Product Description
SAFC
Allyl chloride
Sigma-Aldrich
Allyl chloride, ReagentPlus®, 99%
Sigma-Aldrich
Allyl chloride, reagent grade, 98%