- Effective functionalization of multiwalled carbon nanotube with amphiphilic poly(propyleneimine) dendrimer carrying silver nanoparticles for better dispersability and antimicrobial activity.
Effective functionalization of multiwalled carbon nanotube with amphiphilic poly(propyleneimine) dendrimer carrying silver nanoparticles for better dispersability and antimicrobial activity.
Two multiwalled carbon nanotube hybrids have been prepared: (a) multiwalled carbon nanotubes (MWCNTs) functionalized with amphiphilic poly(propyleneimine) dendrimer (APPI), viz. MWCNTs-APPI, and (b) silver nanoparticles (AgNPs)-deposited multiwalled carbon nanotubes functionalized with an amphiphilic poly(propyleneimine) dendrimer (MWCNTs-APPI-AgNPs). The degree of covalent functionalization of APPI in MWCNTs and deposition of AgNPs in MWCNTs-APPI were examined by Fourier transform infrared spectroscopy, zeta potential, scanning and high-resolution transmission electron microscopy, energy-dispersive spectroscopy, thermogravimetric analysis, and Raman spectroscopy. The amount of APPI functionalized on MWCNTs determined by thermal gravimetric analysis was about 67% which enables an effective dispersability in aqueous and organic solvents without sonication and these solutions were stable for 6 months without undergoing aggregation of MWCNTs. The electronic properties of the hybrid materials were not altered drastically as verified by the Raman studies. The antimicrobial activities of MWCNTs-APPI and MWCNTs-APPI-AgNPs against three different bacteria, viz. Bacillus subtilis, Staphylococcus aureus, and Escheriachia coli illustrated excellent activity.