Merck
Synthesis of trimethylolpropane esters of oleic acid by Lipoprime 50T.
Journal of industrial microbiology & biotechnology (2011-02-18)
Vita Kiriliauskaitė, Vida Bendikienė, Benediktas Juodka
ABSTRACT
The ability of the commercial lipolytic enzyme Lipoprime 50T to catalyze the biotechnologically important synthesis of the biodegradable and environmentally acceptable trimethylolpropane (2-ethyl-2-(hydroxymethyl)-1,3-propanediol) ester of oleic acid was investigated. Simple and accurate thin-layer chromatography and computer analysis methods were used that enable one to follow changes of all reaction mixture components simultaneously. The processes of transesterification and esterification were compared. The effects of the molar ratio of the substrates, reaction temperature, time, and medium on the composition of the reaction mixture were analyzed. Esterification was determined to be more preferable than transesterification in both studied solvents. Under the optimal conditions identified (15% w/w water, temperature 60°C, trimethylolpropane to oleic acid molar ratio 1:3.5, and reaction time 72 h), the highest trimethylolpropane trioleate yield of around 62% and trimethylolpropane mono-, di-, and trioleate overall yield of about 83% were obtained. Although the yields are not high enough for industrial application, the process shows the potential to be optimized for higher yields in the near future as the conversions were obtained at ambient pressure, whereas many other processes described in the literature are conducted under vacuum at a specific pressure.
Sigma-Aldrich
1,1,1-Tris(hydroxymethyl)propane, dist., ≥98.0% (GC)
Sigma-Aldrich
1,1,1-Tris(hydroxymethyl)propane, 97%