Merck
  • Comparison of volatile constituents extracted from model grape juice and model wine by stir bar sorptive extraction-gas chromatography-mass spectrometry.
Comparison of volatile constituents extracted from model grape juice and model wine by stir bar sorptive extraction-gas chromatography-mass spectrometry.
Journal of chromatography. A (2011-01-11)
Darren J Caven-Quantrill, Alan J Buglass
ABSTRACT
A stir bar sorptive extraction (SBSE) method coupled with gas chromatography-mass spectrometry was optimised for the analysis of volatile components of a model wine, based on a previously optimised method used for analysis of the same components in model grape juice. The presence of ethanol in the model wine sample matrix resulted in decreased sensitivity of the method toward most of the volatile constituents. Mean percent relative recoveries and reproducibilities (%CV) were 22.8% and 7.1%, respectively, compared with 28.4% and 8.5% for model grape juice. The mean limit of detection (LoD) ratio (juice:wine) was 0.25. Similar sensitivities for the two sample matrices using this method were achieved by changing the split ratio from 20:1 (grape juice) to 5:1 (wine), giving a mean limit of detection ratio (juice:wine) of 1.0, thus allowing direct comparison of chromatograms of volatile components in the two matrices. This enabled direct comparisons of grape juices and the wines derived from them by alcoholic yeast fermentation. The influence of ethanol concentration in the range 9-15% on method sensitivity is discussed, using an overlay of the total ion chromatograms. The use of a gas saver device for the 5:1 split ratio analysis of desorbed model wine aroma compounds is discussed in terms of preventing extraneous reaction of sorbent and stationary phases with air during analysis.
Sigma-Aldrich
1-Decanol, ≥98%, FCC, FG
Sigma-Aldrich
1-Decanol, 98%
Sigma-Aldrich
1-Decanol, ≥98%
Supelco
1-Decanol, analytical standard