Merck
[Studies on the chemical constituents of Wikstroemia indica].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2010-12-09)
Huan-Kai Yao, Wen-Ting Zhang, Yi-Sang Gao, Ying Zhong
ABSTRACT
To study the chemical constituents of Wikstroemia indica. The constituents were isolated and purified by silica gel chromatography repeatedly, and the structures were determined by spectral data and chemical envidance. Six compounds were isolated from its petroleum ether, dichloromethane, acetone and methanol extracts and identified as: daphnoretin-7-O-beta-D-glucoside (1), aloe-emodin (2), kaempferol (3), 29-nonacosanolide (4), 1-octadecanol (5), beta-sitosterol (6). Compounds 2, 4, 5 are isolated from this plant for the first time.
Product Description
Supelco
1-Octadecanol, Selectophore, ≥99.0%
Sigma-Aldrich
1-Octadecanol, ReagentPlus®, 99%
Sigma-Aldrich
1-Octadecanol, 95%
Supelco
Stearyl Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Stearyl alcohol, European Pharmacopoeia (EP) Reference Standard