Merck
  • Iron-mediated [3 + 2] or [3 + 3] annulation of 2-(2-(ethynyl)phenoxy)-1-arylethanones: selective synthesis of indeno[1,2-c]chromenes and 5H-naphtho[1,2-c]chromenes.
Iron-mediated [3 + 2] or [3 + 3] annulation of 2-(2-(ethynyl)phenoxy)-1-arylethanones: selective synthesis of indeno[1,2-c]chromenes and 5H-naphtho[1,2-c]chromenes.
Organic letters (2010-12-03)
Zhi-Qiang Wang, Yong Lei, Ming-Bo Zhou, Guo-Xiang Chen, Ren-Jie Song, Ye-Xiang Xie, Jin-Heng Li
ABSTRACT
An iron-mediated tandem annulation strategy has been developed for the synthesis of numerous functional indeno[1,2-c]chromenes and 5H-naphtho[1,2-c]chromenes. This work is the first to disclose an iron-mediated method through sequential electrophilic addition of a ketone to an alkyne and annulation tandem reaction. Importantly, a halide is introduced into the products by a ring-opening process among the annulation of alkynylcyclopropanes, which makes the methodology more attractive for organic synthesis.
Sigma-Aldrich
Indene, 98%
Sigma-Aldrich
Indene, contains 50-100 ppm tert-butylcatechol as stabilizer, technical grade, ≥90%
Sigma-Aldrich
Indene, ≥99%