Merck
  • Diphenylthiourea, a common rubber chemical, is bioactivated to potent skin sensitizers.
Diphenylthiourea, a common rubber chemical, is bioactivated to potent skin sensitizers.
Chemical research in toxicology (2010-11-16)
Kristin Samuelsson, Moa Andresen Bergström, Charlotte A Jonsson, Gunnar Westman, Ann-Therese Karlberg
ABSTRACT
Diphenylthiourea (DPTU) is a known skin sensitizer commonly used as a vulcanization accelerator in the production of synthetic rubber, for example, neoprene. The versatile usage of neoprene is due to the multifaceted properties of the material; for example, it is stretchable, waterproof, and chemical- and abrasion-resistant. The wide application of neoprene has resulted in numerous case reports of dermatitis patients allergic to DPTU. The mechanism by which DPTU works as a contact allergen has not been described; thus, the aim of the present study was to investigate if DPTU is a prohapten that can be activated by skin metabolism. The metabolic activation and covalent binding of (14)C-labeled DPTU to proteins were tested using a skinlike cytochrome P450 (P450) cocktail containing the five most abundant P450s found in human skin (CYP1A1, 1B1, 2B6, 2E1, and 3A5) and human liver microsomes. The incubations were carried out in the presence or absence of the metabolite trapping agents glutathione, methoxylamine, and benzylamine. The metabolism mixtures were analyzed by LC-radiochromatography, LC-MS, and LC-MS/MS. DPTU was mainly metabolically activated to reactive sulfoxides resulting in desulfurated adducts in both enzymatic systems used. Also, phenylisothiocyanate and phenylisocyanate were found to be metabolites of DPTU. The sensitizing capacity of the substrate (DPTU) and three metabolites was tested in the murine local lymph node assay. Two out of three metabolites tested were strong skin sensitizers, whereas DPTU itself, as previously known, was negative using this mouse model. In conclusion, DPTU forms highly reactive metabolites upon bioactivation by enzymes present in the skin. These metabolites are able to induce skin sensitization and are probable causes for DPTU allergy. To increase the possibilities of diagnosing contact allergy to DPTU-containing items, we suggest that suitable metabolites of DPTU should be used for screening testing.
Sigma-Aldrich
Benzylamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Methoxyamine hydrochloride, 98%
Sigma-Aldrich
Benzylamine, ReagentPlus®, 99%
Sigma-Aldrich
Phenyl isothiocyanate, for HPLC derivatization, the detection of alcohols and amines, ≥99.0%
Sigma-Aldrich
Phenyl isothiocyanate, reagent grade, 98%
Sigma-Aldrich
Phenyl isothiocyanate, Sigma Grade, 8.36 M, suitable for solid phase protein sequencing analysis, ≥99% (GC), liquid
Sigma-Aldrich
Benzylamine hydrochloride
Supelco
Benzylamine, for GC derivatization, LiChropur, ≥99.0%
Sigma-Aldrich
Phenyl isothiocyanate, 99%, for protein sequencing
Sigma-Aldrich
N,N′-Diphenylthiourea, 98%