Merck
  • Approaches to cancer assessment in EPA's Integrated Risk Information System.
Approaches to cancer assessment in EPA's Integrated Risk Information System.
Toxicology and applied pharmacology (2010-11-03)
Martin W Gehlhaus, Jeffrey S Gift, Karen A Hogan, Leonid Kopylev, Paul M Schlosser, Abdel-Razak Kadry
ABSTRACT
The U.S. Environmental Protection Agency's (EPA) Integrated Risk Information System (IRIS) Program develops assessments of health effects that may result from chronic exposure to chemicals in the environment. The IRIS database contains more than 540 assessments. When supported by available data, IRIS assessments provide quantitative analyses of carcinogenic effects. Since publication of EPA's 2005 Guidelines for Carcinogen Risk Assessment, IRIS cancer assessments have implemented new approaches recommended in these guidelines and expanded the use of complex scientific methods to perform quantitative dose-response assessments. Two case studies of the application of the mode of action framework from the 2005 Cancer Guidelines are presented in this paper. The first is a case study of 1,2,3-trichloropropane, as an example of a chemical with a mutagenic mode of carcinogenic action thus warranting the application of age-dependent adjustment factors for early-life exposure; the second is a case study of ethylene glycol monobutyl ether, as an example of a chemical with a carcinogenic action consistent with a nonlinear extrapolation approach. The use of physiologically based pharmacokinetic (PBPK) modeling to quantify interindividual variability and account for human parameter uncertainty as part of a quantitative cancer assessment is illustrated using a case study involving probabilistic PBPK modeling for dichloromethane. We also discuss statistical issues in assessing trends and model fit for tumor dose-response data, analysis of the combined risk from multiple types of tumors, and application of life-table methods for using human data to derive cancer risk estimates. These issues reflect the complexity and challenges faced in assessing the carcinogenic risks from exposure to environmental chemicals, and provide a view of the current trends in IRIS carcinogenicity risk assessment.
Product Description
Supelco
1,2,3-Trichloropropane, analytical standard
Supelco
1,2,3-Trichloropropane solution, certified reference material, 200 μg/mL in methanol