Merck
  • Merging nucleophilic and hydrogen bonding catalysis: an anion binding approach to the kinetic resolution of propargylic amines.
Merging nucleophilic and hydrogen bonding catalysis: an anion binding approach to the kinetic resolution of propargylic amines.
Journal of the American Chemical Society (2010-09-17)
Eric G Klauber, Chandra Kanta De, Tejas K Shah, Daniel Seidel
ABSTRACT
An efficient kinetic resolution of primary propargylic amines with s-factors of up to 56 is reported. The strategy is based on a dual catalytic approach, namely the use of a newly developed and easy-to-make thiourea-amide anion binding catalyst in combination with 4-(dimethylamino)pyridine (DMAP), both employed at a 5 mol % catalyst loading. Benzylic amines are also resolved with s-factors of up to 38.
Sigma-Aldrich
Propargylamine, 98%
Sigma-Aldrich
4-(Dimethylamino)pyridine, purum, ≥98.0% (NT)
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Propargylamine hydrochloride, 95%