Merck
  • Inhibitory influence of protease-activated receptor 2 and E-prostanoid receptor stimulants in lipopolysaccharide models of acute airway inflammation.
Inhibitory influence of protease-activated receptor 2 and E-prostanoid receptor stimulants in lipopolysaccharide models of acute airway inflammation.
The Journal of pharmacology and experimental therapeutics (2010-08-07)
Terence Peters, Tracy S Mann, Peter J Henry
ABSTRACT
Protease-activated receptors (PARs) are widely expressed throughout the respiratory tract, and PAR(2) has been investigated as a potential drug target for inflammatory airway diseases. The primary focus of this study was to determine the extent to which PAR(2)-activating peptides modulate lipopolysaccharide (LPS)-induced airway neutrophilia in mice and establish the underlying mechanisms. Intranasal administration of LPS induced dose- and time-dependent increases in the number of neutrophils recovered from bronchoalveolar lavage (BAL) fluid of mice. Coadministration of the PAR(2)-activating peptide f-LIGRL inhibited LPS-induced neutrophilia at 3 and 6 h after inoculation. PAR(2)-mediated inhibition of LPS-induced neutrophilia was mimicked by prostaglandin E(2) (PGE(2)) and butaprost [selective E-prostanoid (EP(2)) receptor agonist], and blocked by parecoxib (cyclooxygenase 2 inhibitor) and 6-isopropoxy-9-oxoxanthene-2-carboxylic acid (AH6809) (EP(1)/EP(2) receptor antagonist). PAR(2)-activating peptides also blunted early increases in the levels of the key neutrophil chemoattractants keratinocyte-derived chemokine and macrophage inflammatory protein 2 (MIP-2) in the BAL of LPS-exposed mice. However, neither PAR(2)-activating peptides nor PGE(2) inhibited LPS-induced generation of MIP-2 in cultures of primary murine alveolar macrophages In summary, PAR(2)-activating peptides and PGE(2) suppressed LPS-induced neutrophilia in murine airways, independently of an inhibitory action on MIP-2 generation by alveolar macrophages.
Sigma-Aldrich
(R)-Butaprost, ≥98% (HPLC)