Merck
  • Synthesis and bioactivity of N-benzoyl-N'-[5-(2'-substituted phenyl)-2-furoyl] semicarbazide derivatives.
Synthesis and bioactivity of N-benzoyl-N'-[5-(2'-substituted phenyl)-2-furoyl] semicarbazide derivatives.
Molecules (Basel, Switzerland) (2010-07-27)
Zining Cui, Yun Ling, Baoju Li, Yongqiang Li, Changhui Rui, Jingrong Cui, Yanxia Shi, Xinling Yang
ABSTRACT
In order to find novel chitin synthesis inhibitors (CSIs) with good activity, benzoylphenylurea, a typical kind of CSIs, was chosen as the lead compound and 15 novel derivatives containing furan moieties were designed by converting the urea linkage of benzoylphenylureas into a semicarbazide and changing the aniline part into furoyl groups. The title compounds were synthesized by the reaction of substituted benzoyl isocyanates with 5-(substituted phenyl)-2-furoyl hydrazine, and the structures were confirmed by IR, (1)H-NMR, elemental analysis and single crystal X-ray diffraction analyses (compound E2). The bioassay results indicated that the title compounds exhibit good insecticidal activity, especially towards Plutella xylostella L., but had lower fungicidal activity. Inspiringly, the title compounds possessed obvious anticancer activity against human promyelocytic leukemic cell line (HL-60), and some of the title compounds also had activity against human hepatocellular carcinoma cell line (Bel-7402), human gastric carcinoma cell line (BGC-823), and human nasopharyngeal carcinoma cell line (KB). The results indicated that the linkage in the lead compounds was important to the bioactivity and spectra. The modification on the urea linkage is an effective strategy to discover new pesticide and drug candidates.
Product Description
Supelco
Semicarbazide hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Semicarbazide hydrochloride, ≥99%
Sigma-Aldrich
Semicarbazide, 6 wt. % (on silica gel)