Merck
  • Induction of hexokinase I expression in normal and diabetic rats by a brassinosteroid isoform.
Induction of hexokinase I expression in normal and diabetic rats by a brassinosteroid isoform.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2010-06-24)
P Muthuraman, K Srikumar
ABSTRACT
This study aimed to investigate the effect of the plant growth regulator 28-homobrassinolide (HB) on the hexokinase I (HK I) enzyme gene expression in the tissues of normal and streptozotocin-induced diabetic rats. Normal and diabetic rats were administered 50 microg of HB for 15 consecutive days. The tissues level of HK I mRNA expression was quantitated by PCR and densitometry analysis, HK I protein expression was quantitated by Western blot and densitometry analysis, localization of HK I was done by immunohistochemistry and HK enzyme activity was determined by coupled enzyme assay. Subchronic treatment of rats with HB enhanced HK I enzyme expression in diabetic rat compared to the control rat. From these experimental evidences, we came to the conclusion that HK I played a vital role in the regulation of blood sugar in streptozotocin-induced diabetic rats. A direct role for hexokinase enzyme activity in the control of diabetes is presented.
Sigma-Aldrich
22(S),23(S)-Homobrassinolide