Merck
  • Influence of short chain organic acids and bases on the wetting properties and surface energy of submicrometer ceramic powders.
Influence of short chain organic acids and bases on the wetting properties and surface energy of submicrometer ceramic powders.
Journal of colloid and interface science (2010-06-18)
Bram Neirinck, Dimitri Soccol, Jan Fransaer, Omer Van der Biest, Jef Vleugels
ABSTRACT
The effect of short chained organic acids and bases on the surface energy and wetting properties of submicrometer alumina powder was assessed. The surface chemistry of treated powders was determined by means of Diffuse Reflectance Infrared Fourier Transform spectroscopy and compared to untreated powder. The wetting of powders was measured using a modified Washburn method, based on the use of precompacted powder samples. The geometric factor needed to calculate the contact angle was derived from measurements of the porous properties of the powder compacts. Contact angle measurements with several probe liquids before and after modification allowed a theoretical estimation of the surface energy based on the surface tension component theory. Trends in the surface energy components were linked to observations in infrared spectra. The results showed that the hydrophobic character of the precompacted powder depends on both the chain length and polar group of the modifying agent.
Sigma-Aldrich
Butylamine, ≥99%
Sigma-Aldrich
Butylamine, 99.5%
Supelco
Valeric acid, analytical standard
Sigma-Aldrich
Valeric acid, ≥99%
Sigma-Aldrich
Valeric acid, ≥99%, FCC, FG