Merck
  • Formation of a quinoneimine intermediate of 4-fluoro-N-methylaniline by FMO1: carbon oxidation plus defluorination.
Formation of a quinoneimine intermediate of 4-fluoro-N-methylaniline by FMO1: carbon oxidation plus defluorination.
Chemical research in toxicology (2010-04-08)
James P Driscoll, Ignacio Aliagas, Jennifer J Harris, Jason S Halladay, Sheerin Khatib-Shahidi, Alan Deese, Nathaniel Segraves, S Cyrus Khojasteh-Bakht
ABSTRACT
Here, we report on the mechanism by which flavin-containing monooxygenase 1 (FMO1) mediates the formation of a reactive intermediate of 4-fluoro-N-methylaniline. FMO1 catalyzed a carbon oxidation reaction coupled with defluorination that led to the formation of 4-N-methylaminophenol, which was a reaction first reported by Boersma et al. (Boersma et al. (1993) Drug Metab. Dispos. 21 , 218 - 230). We propose that a labile 1-fluoro-4-(methylimino)cyclohexa-2,5-dienol intermediate was formed leading to an electrophilic quinoneimine intermediate. The identification of N-acetylcysteine adducts by LC-MS/MS and NMR further supports the formation of a quinoneimine intermediate. Incubations containing stable labeled oxygen (H(2)(18)O or (18)O(2)) and ab initio calculations were performed to support the proposed reaction mechanism.
Sigma-Aldrich
4-Fluoro-N-methylaniline, 97%