Merck
Amino acid salt catalyzed intramolecular Robinson annulation.
Chemical communications (Cambridge, England) (2009-09-03)
Pingfan Li, Hisashi Yamamoto
ABSTRACT
The silica gel absorbed amino acid salt catalyzed asymmetric intramolecular Robinson annulation reaction has been developed; up to 97% ee was obtained with this readily recoverable organocatalyst.
Sigma-Aldrich
1,4-Cyclohexanedimethanol, mixture of cis and trans, 99%