Merck
  • [Olfactory functional magnetic resonance imaging with modified OEP-98C olfactometer and event-related design].
[Olfactory functional magnetic resonance imaging with modified OEP-98C olfactometer and event-related design].
Zhongguo yi xue ke xue yuan xue bao. Acta Academiae Medicinae Sinicae (2009-06-11)
Hui You, Jian Wang, Jian-Feng Liu, Feng Feng, Dao-Feng Ni, Zheng-Yu Jin
ABSTRACT
To explore the feasibility of functional magnetic resonance imaging (fMRI) in analysis of olfaction function with modified OEP-98C olfactometer and event-related design. Six young right-handed men underwent olfactory fMRI with event-related design. OEP-98C olfactometer was modified to accommodate MR environment. There were 2 types of tasks in the experiment. In one task, only isoamyl acetate was used as odorant. In the other task, to avoid possible decreased olfactory attention, vanillin was given before each presentation of isoamyl acetate. In both tasks, uniform activation in piriform cortex and secondary olfactory cortexes was determined. The activation of piriform cortex was not significantly different between the two tasks (P > 0.01). With isoamyl acetate as odorant, modified OEP-98C olfactometer, and event-related design, olfaction fMRI can depict cortex activation at primary and secondary olfactory cortex. Applying other odorant with similar quality to avoid olfactory attention decrease can not promote depiction of activation in primary olfactory cortex.
Sigma-Aldrich
Isopentyl acetate, anhydrous, ≥99%
Supelco
Isopentyl acetate, analytical standard
Sigma-Aldrich
Isopentyl acetate, mixture of isomers, reagent grade, 98%
Sigma-Aldrich
Isoamyl acetate, ≥95%, FCC, FG
Sigma-Aldrich
Isoamyl acetate, natural, ≥97%, FCC, FG