Merck
  • Selective monomethylation of anilines by Cu(OAc)2-promoted cross-coupling with MeB(OH)2.
Selective monomethylation of anilines by Cu(OAc)2-promoted cross-coupling with MeB(OH)2.
Organic letters (2009-04-10)
Israel González, Jesús Mosquera, César Guerrero, Ramón Rodríguez, Jacobo Cruces
ABSTRACT
N-Methylanilines are readily synthesized in high yields through the copper(II)-promoted coupling of anilines and methylboronic acid. This method represents a new approach for the selective monomethylation of anilines, and it is the first reported example of a Chan-Lam coupling involving the use of methylboronic acid. An incubation period of the substrate with the copper reagent is needed before addition of the methylboronic acid.
Sigma-Aldrich
N-Methylaniline, purum, ≥98.0% (GC)
Supelco
N-Methylaniline, analytical standard
Sigma-Aldrich
N-Methylaniline, ≥99%
Sigma-Aldrich
N-Methylaniline, 98%