Merck
Fate of benfuracarb insecticide in mollisols and brinjal crop.
Bulletin of environmental contamination and toxicology (2009-04-01)
Raju Chandra, Anjana Srivastava, Prakash C Srivastava
ABSTRACT
The fate of benfuracarb was studied under field conditions in brinjal fruits and soil following foliar spray application at 0.25 and 0.50 microg g(-1) by HPLC. At 0.25 microg g(-1), benfuracarb persisted up to 7 days both in soil and brinjal but at 0.50 microg g(-1), benfuracarb residues persisted up to 10 and 12 days in soil and brinjal fruits, respectively. The persistence of benfuracarb residues, both in soil and brinjal, followed first-order kinetics. The half-life values of benfuracarb in soil and brinjal fruit were found to be 3.54 and 3.90 days at 0.25 microg g(-1) and 3.75 and 4.73 days at 0.50 microg g(-1), respectively.
Product Description
Supelco
Benfuracarb, PESTANAL®, analytical standard