Merck
  • Development of redox-labeled electrochemical immunoassay for polycyclic aromatic hydrocarbons with controlled surface modification and catalytic voltammetric detection.
Development of redox-labeled electrochemical immunoassay for polycyclic aromatic hydrocarbons with controlled surface modification and catalytic voltammetric detection.
Biosensors & bioelectronics (2009-03-27)
Ming-Yuan Wei, Shou-Dong Wen, Xi-Qiang Yang, Liang-Hong Guo
ABSTRACT
A redox-labeled direct competitive electrochemical immunoassay for polycyclic aromatic hydrocarbons (PAHs) was developed. A ruthenium tris(bipyridine)-pyrenebutyric acid conjugate was synthesized as the redox-labeled tracer. Its recognition by an anti-PAH monoclonal antibody was confirmed by surface plasmon resonance. In the immunoassay, the antibody was immobilized on (3-glycidoxypropyl)-trimethoxysilane (GPTMS)-modified indium tin oxide (ITO) electrodes. The assay was quantified by measuring the electro-catalytic current of the redox label in an oxalate-containing electrolyte which served as a sacrificial electron donor to amplify the current signal. Formation of GPTMS film on ITO and subsequent antibody immobilization were characterized by X-ray photoelectron spectroscopy (XPS) and electrochemistry. Using a ruthenium tris(bipyridine)-conjugated IgG (IgG-Ru) as the surface-bound redox probe, the highest electrochemical signal was obtained on GPTMS electrodes with 1 h modification. Under the optimized conditions for ITO modification, antibody immobilization and tracer concentration, competition curves for benzo[a]pyrene and pyrenebutyric acid were obtained with a detection limit of 2.4 and 10 ng mL(-1), respectively. The redox-labeled electrochemical immunoassay with signal amplification mechanism offers a potential analytical method for the simultaneous detection of multiple environmental organic pollutants on antibody biochips.
Sigma-Aldrich
1-Pyrenebutyric acid, 97%