Merck
  • Inhibition of methanogens by bromochloromethane: effects on microbial communities and rumen fermentation using batch and continuous fermentations.
Inhibition of methanogens by bromochloromethane: effects on microbial communities and rumen fermentation using batch and continuous fermentations.
The British journal of nutrition (2009-02-27)
Gunjan Goel, Harinder P S Makkar, Klaus Becker
ABSTRACT
Bromochloromethane (BCM), a halogenated methane analogue, was evaluated for its anti-methanogenic activity in both batch and continuous fermentation systems. For batch fermentation, a roughage-based substrate was incubated with mixed rumen microflora for 24 h at two concentrations of BCM (5 and 10 microm). A 89-94 % reduction in methane was obtained in terms of volume and truly degraded substrate (TDS; P < 0.05). The partitioning factor (an index of efficiency of microbial protein production; expressed as mg TDS/ml net gas produced) increased from 3.55 to 3.73 (P < 0.05), while the acetate:propionate proportion decreased from 2.80 to 2.22 (P < 0.05). A complete inhibition of methanogens was associated with a 48 % decrease in Ruminococcus flavefaciens, a 68 % increase in Fibrobacter succinogenes and a 30 % increase in rumen fungi when quantified using qPCR. The continuous fermentation was carried out using the roughage-based substrate in four fermenters, two fermenters being control and the other two fed with BCM (5 microm) once in a day, for nine consecutive days. A persistent effect of BCM on methane reduction (85-90 %) was obtained throughout the study (P < 0.05) with no effect on gas production, SCFA production, acetate:propionate proportion, true degradability and efficiency of microbial mass synthesis (P>0.05). The complete inhibition of methane resulted in a significant decrease in R. flavefaciens and methanogens (P < 0.05) and an increase in fungal population (P < 0.05), while there was no effect on total bacterial and F. succinogenes populations (P>0.05). The batch fermentation confirms the anti-methanogenic activity of BCM, while the continuous fermentation indicates the persistency of this effect under in vitro conditions.
Sigma-Aldrich
Bromochloromethane, contains 100 ppm BHT as inhibitor, ≥99.5%
Supelco
Bromochloromethane, analytical standard