Merck
  • A diversity oriented, microwave assisted synthesis of N-substituted 2-hydro-4-amino-pyrido[2,3-d]pyrimidin-7(8H)-ones.
A diversity oriented, microwave assisted synthesis of N-substituted 2-hydro-4-amino-pyrido[2,3-d]pyrimidin-7(8H)-ones.
Molecular diversity (2008-11-28)
Núria Mont, Jordi Teixidó, José I Borrell
ABSTRACT
A protocol for the synthesis of N-substituted 2-hydro-4-amino-pyrido[2,3-d]pyrimidin-7(8H)-ones (11) is described. Thus, the formylation of a 2-aminopyridone 12 in 85% formic acid/Ac(2)O, proceeding via in situ cyclization to the intermediate formamide 13, affords the corresponding 2-hydro-4-oxo-pyridopyrimidine 14, which is converted to a 4-chloro-pyridopyrimidine 15 upon treatment with POCl(3). The subsequent transformation to the title compounds is carried by treatment with the corresponding amine in MeOH under microwave irradiation conditions.
Sigma-Aldrich
Phosphorus trichloride, ReagentPlus®, 99%
Sigma-Aldrich
Phosphorus trichloride, 99.999% trace metals basis