Merck
  • Molecularly imprinted polymer of 5-methyluridine for solid-phase extraction of pyrimidine nucleoside cancer markers in urine.
Molecularly imprinted polymer of 5-methyluridine for solid-phase extraction of pyrimidine nucleoside cancer markers in urine.
Bioorganic & medicinal chemistry (2008-09-16)
Damien Jégourel, Raphaël Delépée, Florent Breton, Antoine Rolland, Richard Vidal, Luigi A Agrofoglio
ABSTRACT
Normal and modified urinary nucleosides represent potential biomarkers for cancer diagnosis. To selectively extract modified nucleosides, we developed a molecularly imprinted polymer (MIP) of 5-methyluridine as selective material for molecularly imprinted solid-phase extraction (MISPE). The MIPs were obtained from vinyl-phenylboronate ester derivative of the template, acrylamide and pentaerythritol triacrylate co-polymer, and were tested in batch and cartridge experiments with aqueous samples. Our results indicated that the imprinted polymer was selective for pyrimidine nucleosides with a K(d) and a B(max) of 46 microM and 18 micromol/g, respectively. Finally, a MISPE of the most common pyrimidine nucleoside cancer markers in urine sample was realized.
Sigma-Aldrich
Pentaerythritol tetraacrylate, Pentaerythritol triacrylate, and Trimethylolpropane triacrylate mixture
Sigma-Aldrich
5-Methyluridine, 97%