Merck
  • Concurrent anion...pi interactions between a perchlorate ion and two pi-acidic aromatic rings, namely pentafluorophenol and 1,3,5-triazine.
Concurrent anion...pi interactions between a perchlorate ion and two pi-acidic aromatic rings, namely pentafluorophenol and 1,3,5-triazine.
Chemical communications (Cambridge, England) (2008-07-18)
Rens J Götz, Arturo Robertazzi, Ilpo Mutikainen, Urho Turpeinen, Patrick Gamez, Jan Reedijk
ABSTRACT
The rational design of a ligand containing two electron-poor pi-rings, i.e. a triazine and a pentafluorophenoxy groups, has allowed the preparation of a copper complex where both the anticipated anion...pi interactions are present.
Sigma-Aldrich
Pentafluorophenol, ReagentPlus®, ≥99%