Merck
Occurrence of 3-MCPD fatty acid esters in human breast milk.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2008-05-20)
Z Zelinková, O Novotný, J Schůrek, J Velísek, J Hajslová, M Dolezal
ABSTRACT
A series of twelve breast milk samples were analysed by gas chromatography-mass spectrometry (GC/MS) operated in selected ion monitoring mode for 3-chloropropane-1,2-diol (3-MCPD). Whilst none of the samples contained 3-MCPD above the limit of detection of 3 microg kg(-1) milk, all contained high amounts of 3-MCPD esterified with higher fatty acids. The levels of 3-MCPD released by hydrolysis of these esters (bound 3-MCPD) ranged from the limit of detection (300 microg kg(-1), expressed on a fat basis) to 2195 microg kg(-1); with a mean level of bound 3-MCPD of 1014 microg kg(-1), which corresponded to 35.5 microg kg(-1) milk. The presence of bound 3-MCPD was confirmed using orthogonal gas chromatography coupled with high-speed time-of-flight mass spectrometry analysis for four randomly selected breast milk samples. Six breast milks collected from one of the nursing mothers 14-76 days after childbirth contained bound 3-MCPD within the range of 328-2078 microg kg(-1) fat (mean 930 microg kg(-1) fat). The calculated bound 3-MCPD content of these samples was within the range of 6 and 19 microg kg(-1) milk (mean of 12 microg kg(-1) milk). The major types of 3-MCPD esters were the symmetric diesters with lauric, palmitic, and oleic acids, and asymmetric diesters with palmitic acid/oleic acid among which 3-chloro-1,2-propanediol 1,2-dioleate prevailed.
Sigma-Aldrich
(R)-(−)-3-Chloro-1,2-propanediol, 97%, optical purity ee: 98% (GLC)
Sigma-Aldrich
(S)-(+)-3-Chloro-1,2-propanediol, 97%, optical purity ee: 97% (GLC)
Sigma-Aldrich
(±)-3-Chloro-1,2-propanediol, 98%
Supelco
(±)-3-Chloro-1,2-propanediol, analytical standard