Merck
Effects of cuminaldehyde on melanoma cells.
Phytotherapy research : PTR (2008-04-17)
Teruhiko Nitoda, Maria D Fan, Isao Kubo
ABSTRACT
Cuminaldehyde (4-isopropylbenzaldehyde) suppressed melanin formation in cultured murine B16-F10 melanoma cells in a dose-dependent decrease up to 0.25 mm without affecting cell growth. Approximately 30% suppression in melanin production resulted when the cells were cultured with 0.25 mm of cuminaldehyde. This activity was not noticeable with cultured human A375 melanoma cells.
Sigma-Aldrich
Cuminaldehyde, ≥98%, FCC, FG
Sigma-Aldrich
Cuminaldehyde, 98%