Merck
  • Development and validation of HPLC method for the resolution of drug intermediates: DL-3-Phenyllactic acid, DL-O-acetyl-3-phenyllactic acid and (+/-)-mexiletine acetamide enantiomers.
Development and validation of HPLC method for the resolution of drug intermediates: DL-3-Phenyllactic acid, DL-O-acetyl-3-phenyllactic acid and (+/-)-mexiletine acetamide enantiomers.
Talanta (2008-03-29)
Alemu Tekewe, Sawraj Singh, Manpreet Singh, Utpal Mohan, U C Banerjee
ABSTRACT
Sensitive and specific, high-performance liquid chromatography (HPLC) methods have been developed and validated for linearity, accuracy and precision for the quantification of dl-3-phenyllactic acid, dl-O-acetyl-3-phenyllactic acid and (+/-)-mexiletine acetamide enantiomers. Chromatographic separations were performed on a Chiralcel OJ-H column (0.46 mm x 250 mm, 5 microm, Daicel Chemical Industries, Japan) based on cellulose tris-(4-methyl benzoate) chiral stationary phase. The mobile phase consists of hexane and isopropanol (IPA) in the ratio of 90:10 containing 0.1% trifluoroacetic acid (in case of dl-3-phenyllactic acid and dl-O-acetyl-3-phenyllactic acid) and hexane and IPA (95:5) containing 0.1% triethylamine (in case of (+/-)-mexiletine acetamide) and the flow rate was set at 0.5 ml/min at 25 degrees C. The detection was carried out at 261 nm for dl-3-phenyllactic acid and dl-O-acetyl-3-phenyllactic acid and at 254 nm for (+/-)-mexiletine acetamide. The developed methods were utilized for monitoring the progress of lipase catalyzed enantioselective synthesis of O-acetyl-3-phenyllactic acid and mexiletine acetamide from dl-3-phenyllactic acid and (+/-)-mexiletine, respectively.
Sigma-Aldrich
3-Phenyllactic acid, ≥98%
Sigma-Aldrich
D-(+)-3-Phenyllactic acid, 98%
Sigma-Aldrich
L-(−)-3-Phenyllactic acid, 98%