Merck
  • HPLC determination of serotonin and its metabolites from human platelet-rich plasma; shift to 5-hydroxytryptophol formation following alcohol consumption.
HPLC determination of serotonin and its metabolites from human platelet-rich plasma; shift to 5-hydroxytryptophol formation following alcohol consumption.
Journal of chromatographic science (2008-03-28)
Kornélia Tekes
ABSTRACT
A sensitive, simple, and reliable high-performance liquid chromatographic method with electrochemical detection is developed for the measurement of four natural products, the serotonin-related indols from human platelet-rich plasma (PRP) using N-methylserotonin as internal standard. Separation of serotonin (5HT), 5-hydroxytryptophan (5HTP), 5-hydroxytryptophol (5HTOL), and 5-hydroxyindole-acetic acid (5HIAA) is carried out on Supelcosil LC-18DB stationary phase. A mixture of 48 mM citric acid, 28 mM sodium phosphate dibasic, 0.027 mM Na2EDTA, and 3% methanol (pH 3.18) serves as the mobile phase. Measurements are carried out at 25 degrees C at Eox=0.65 V. The calibration curves are linear through the range of 10-200 pg/mL. Method validation is performed according to internationally accepted criteria. Blood is collected from healthy controls and schizophrenic subjects. Significantly higher PRP serotonin is measured in schizophrenics; patients with recent alcohol consumption could be characterized with significantly elevated 5HTOL/5HIAA ratio.
Sigma-Aldrich
5-Hydroxy--methyltryptamine oxalate, 99%
Pricing and availability is not currently available.
Supelco
SUPELCOSIL LC-NH2-NP HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Pricing and availability is not currently available.
Supelco
SUPELCOSIL LC-NH2-NP Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Pricing and availability is not currently available.
Sigma-Aldrich
Nω-Methyl-5-hydroxy­trypt­amine oxalate salt, ≥90% (HPLC)
Pricing and availability is not currently available.