Merck
  • Simvastatin inhibits growth via apoptosis and the induction of cell cycle arrest in human melanoma cells.
Simvastatin inhibits growth via apoptosis and the induction of cell cycle arrest in human melanoma cells.
Melanoma research (2008-03-14)
Akira Saito, Noriko Saito, William Mol, Hiroshi Furukawa, Arata Tsutsumida, Akihiko Oyama, Mitsuru Sekido, Satoru Sasaki, Yuhei Yamamoto
ABSTRACT
Competitive inhibitors of 3-hydroxy-3-methylglutaryl coenzyme A reductase (the statins) that inhibit the synthesis of mevalonic acid are in wide use for treatment of hypercholesterolemia. Although antitumor effects on a variety of cell types have been reported for statins, the effect of simvastatin (one of the statins) on human melanoma cell lines is not known. Here, we report antitumor effects of simvastatin on human melanoma cell lines. We treated human melanoma cell lines, A375M, G361, C8161, GAK, and MMAc with simvastatin in various concentrations for 1 to 3 days. To investigate the antitumor effect of simvastatin, we analyzed cell viability, morphologic changes, reversibility of inhibition by geranylgeranyl pyrophosphate and farnesyl pyrophosphate, apoptosis and the cell cycle. Simvastatin treatment reduced cell viability in all five melanoma cell lines. The different melanoma cell lines, however, displayed different sensitivities to simvastatin. The addition of geranylgeranyl pyrophosphate to A375M and G361 cells in the presence of simvastatin completely restored the viability of cells, but the addition of farnesyl pyrophosphate did not. DNA fragmentation assay showed that simvastatin induced apoptosis in A375M and G361 cells. Simvastatin caused a G1 arrest in G361 and MMAc cells. Consistent with the cell cycle arrest, simvastatin caused an increase in the mRNA levels of p21 and p27 on G361 and MMAc cells. We conclude that simvastatin has an antitumor effect on human melanoma cells in vitro via apoptosis and cell cycle arrest. These results suggest that simvastatin may be an effective anticancer drug for malignant melanoma.
Product Description
Supelco
Geranyl pyrophosphate lithium salt, analytical standard
Sigma-Aldrich
Geranyl pyrophosphate lithium salt, ≥95.0% (TLC)
Sigma-Aldrich
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)