Merck
  • Chemo-enzymatic supported synthesis of the 3-sulfated Lewis a pentasaccharide on a multimeric polyethylene glycol.
Chemo-enzymatic supported synthesis of the 3-sulfated Lewis a pentasaccharide on a multimeric polyethylene glycol.
Carbohydrate research (2008-02-19)
Annie Malleron, Christine Le Narvor
ABSTRACT
The 3-sulfated Lewis(a) pentasaccharide was synthesized on multimeric-based polyethylene glycol support. Coupling of O-(2,3,4,6-tetra-O-acetyl-beta-D-galactopyranosyl)-(1-->3)-4,6-di-O-acetyl-2-deoxy-2-phthalimido-beta-D-glucopyranosyl trichloroacetimidate with (2,6-di-O-acetyl-beta-D-galactopyranosyl)-(1-->4)-(2,3,6-tri-O-acetyl-beta-D-glucopyranoside) bound onto the polymer afforded lacto-N-tetraose, which was then regioselectively sulfated at the 3-OH position of the terminal galactose using the stannylene procedure. Fucosylation of the sulfated tetrasaccharide was performed using an immobilized fucosyltransferase FucTIII to give the title compound after cleavage.
Sigma-Aldrich
Pentaerythritol, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Please contact Customer Service for Availability
CA$ 28,30
5 g
Please contact Customer Service for Availability
CA$ 86,40
25 g
Please contact Customer Service for Availability
CA$ 247,00
Sigma-Aldrich
Pentaerythritol, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Please contact Customer Service for Availability
CA$ 28,30
5 g
Please contact Customer Service for Availability
CA$ 86,40
25 g
Please contact Customer Service for Availability
CA$ 247,00
Sigma-Aldrich
Guanosine 5′-diphospho-β-L-fucose sodium salt, ≥85%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Please contact Customer Service for Availability
CA$ 28,30
5 g
Please contact Customer Service for Availability
CA$ 86,40
25 g
Please contact Customer Service for Availability
CA$ 247,00