Merck
  • Molecular switching in the near infrared (NIR) to visible/NIR f-f emission with a functional-lanthanide complexes.
Molecular switching in the near infrared (NIR) to visible/NIR f-f emission with a functional-lanthanide complexes.
Journal of fluorescence (2008-02-13)
Ga-Lai Law, Ka-Leung Wong, Yang-Yi Yang, Hai-Ling Yang, Wing-Tak Wong, Michael Hon-Wah Lam, Hoi-Lam Tam, Kok-Wai Cheah
ABSTRACT
We report the first observation of three photon induced NIR emission (890-1,400 nm) with NIR excitation (800 nm) from a newly synthesized organic neodymium complex with known molecular structure, suggests the potential use of lanthanide probes for use in three-dimensional imaging and further the study of multi- photon induced emission process in organic-lanthanide complexes.
Sigma-Aldrich
Terbium(III) chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Terbium(III) chloride hexahydrate, 99.9% trace metals basis
Sigma-Aldrich
Terbium(III) chloride hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Terbium(III) chloride, anhydrous, powder, 99.9% trace metals basis
Sigma-Aldrich
Neodymium(III) chloride, anhydrous, powder, ≥99.99% trace metals basis